ContactUsLinkComponent
EDN2 contains 21 amino acids having a molecular mass of 2546.97 Dalton. 1 * 5 mg
Catalog # BIRBORB80709-5
Supplier:  Biorbyt
EDN2 contains 21 amino acids having a molecular mass of 2546.97 Dalton. 1 * 5 mg
Catalog # BIRBORB80709-5
Supplier:  Biorbyt