TNF -b His Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 171 amino acids fragment (35-205) and having a molecular mass of 23.3kDa with an amino-terminal hexahistidine tag. The TNF-b His is 1 * 20 µG
BIRBORB80621-20
: Biorbyt