ContactUsLinkComponent
TNF-b Human Recombinant (Lymphotoxin) produced in E.coli is a single, non-glycosylated, polypeptide chain containing 172 amino acids and having a molecular mass of 18645 Dalton. The TNF-b is purified by standard chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80620-5
Supplier:  Biorbyt
TNF-b Human Recombinant (Lymphotoxin) produced in E.coli is a single, non-glycosylated, polypeptide chain containing 172 amino acids and having a molecular mass of 18645 Dalton. The TNF-b is purified by standard chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80620-5
Supplier:  Biorbyt