ContactUsLinkComponent
TGF-b Human Recombinant produced in CHO is disulfide-linked homodimeric, glycosylated, polypeptide chain containing 112 amino acids and having a molecular mass of 25kDa. The TGF-b is purified by standard chromatographic techniques. 1 * 100 µG
Catalog # BIRBORB80614-100
Supplier:  Biorbyt
TGF-b Human Recombinant produced in CHO is disulfide-linked homodimeric, glycosylated, polypeptide chain containing 112 amino acids and having a molecular mass of 25kDa. The TGF-b is purified by standard chromatographic techniques. 1 * 100 µG
Catalog # BIRBORB80614-100
Supplier:  Biorbyt