TGF-b3 Human Recombinant produced in E.coli is disulfide-linked homodimeric, non-glycosylated, polypeptide chain containing 112 amin o acids and having a molecular mass of 25,445 Dalton. The TGF-b is purified by standard chromatographic tech 1 * 100 µG
Catalog # BIRBORB80613-100
Supplier: Biorbyt