Human IL5 Recombinant produced in E.coli is dimeric, non-glycosylated polypeptide chain containing 132 amino acids and having a mole cular mass of 26522.84 Dalton.The IL-5 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80425-10
Supplier: Biorbyt