ContactUsLinkComponent
Human IL3 Recombinant produced in insect cells is a single, glycosylated polypeptide chain containing 133 amino acids and having a m olecular mass of 15000 Dalton. The IL3 CSF is fused to C-terminal His-tag (6x His) and purified by proprieta 1 * 2 µG
Catalog # BIRBORB80420-2
Supplier:  Biorbyt
Human IL3 Recombinant produced in insect cells is a single, glycosylated polypeptide chain containing 133 amino acids and having a m olecular mass of 15000 Dalton. The IL3 CSF is fused to C-terminal His-tag (6x His) and purified by proprieta 1 * 2 µG
Catalog # BIRBORB80420-2
Supplier:  Biorbyt