ContactUsLinkComponent
TGFB1 Human Recombinant produced in Human 293 cells is homodimeric polypeptide chain containing x 112 amino acids and having total m olecular mass of 25kDa. The TGFB1 is purified by proprietary chromatographic techniques. 1 * 1 µG
Catalog # BIRBORB80607-1
Supplier:  Biorbyt
TGFB1 Human Recombinant produced in Human 293 cells is homodimeric polypeptide chain containing x 112 amino acids and having total m olecular mass of 25kDa. The TGFB1 is purified by proprietary chromatographic techniques. 1 * 1 µG
Catalog # BIRBORB80607-1
Supplier:  Biorbyt