SF20 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 162 amino acids fragment (33-1 73) and having total molecular mass of 18 kDa. C9orf10 is fused to 20 amino acids His tag at N-terminus and 1 * 10 µG
Catalog # BIRBORB80602-10
Supplier: Biorbyt