IL2 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 170 amino acids (21-169 a.a) and having a molecular mass of 19.5kDa. The mouse IL-2 is fused to 20 amino acid His tag at N-Terminus and puri 1 * 10 µG
Catalog # BIRBORB80414-10
Supplier: Biorbyt