ContactUsLinkComponent
IL1b Porcine Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 154 amino acids and having a molecular mass of 17690 Dalton. The IL-1b is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80409-2
Supplier:  Biorbyt
IL1b Porcine Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 154 amino acids and having a molecular mass of 17690 Dalton. The IL-1b is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80409-2
Supplier:  Biorbyt