IL1b Porcine Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 154 amino acids and having a molecular mass of 17690 Dalton. The IL-1b is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80409-10
Supplier: Biorbyt