ContactUsLinkComponent
IL1 beta Mouse Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 189 amino acids and having a molecul ar mass of 21 kDa. The IL-1b is fused to His-Tag and purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80408-5
Supplier:  Biorbyt
IL1 beta Mouse Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 189 amino acids and having a molecul ar mass of 21 kDa. The IL-1b is fused to His-Tag and purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80408-5
Supplier:  Biorbyt