ContactUsLinkComponent
Soluble FGFR-3a (IIIc) Fc Chimera Human Recombinant fused with Xa cleavage site with the Fc part of human IgG1 produced in baculovir us is heterodimeric, glycosylated, Polypeptide chain and having a molecular mass of 190 kDa. The FGFR3 is pu 1 * 2 µG
Catalog # BIRBORB80805-2
Supplier:  Biorbyt
Soluble FGFR-3a (IIIc) Fc Chimera Human Recombinant fused with Xa cleavage site with the Fc part of human IgG1 produced in baculovir us is heterodimeric, glycosylated, Polypeptide chain and having a molecular mass of 190 kDa. The FGFR3 is pu 1 * 2 µG
Catalog # BIRBORB80805-2
Supplier:  Biorbyt