Soluble FGFR-2a (IIIc) Fc Chimera Human Recombinant fused with Xa cleavage site with the Fc part of human IgG1 produced in baculovir us is heterodimeric, glycosylated, Polypeptide chain and having a molecular mass of 195 kDa. The FGFR2 is pu 1 * 2 µG
Catalog # BIRBORB80804-2
Supplier: Biorbyt