Soluble FGFR-1a (IIIc) Fc Chimera Human Recombinant fused with Xa cleavage site with the Fc part of human IgG1 produced in baculovir us is heterodimeric, glycosylated, Polypeptide chain and having a molecular mass of 170 kDa. The FGFR1 is pu 1 * 2 µG
Catalog # BIRBORB80803-2
Supplier: Biorbyt