Active Focal Adhesion Kinase full length Human Recombinant is 146.7 kDa enzyme produced in Sf9 cells with an amino-terminal GST tag and purified via sequential chromatography. PTK2 is activated in-vitro by his tagged SRC. 1 * 10 µG
Catalog # BIRBORB80802-10
Supplier: Biorbyt