ContactUsLinkComponent
IGFBP6 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing amino acids 148-240 and havi ng a molecular mass of 20 kDa including 4kDa His tag. IGFBP6 is purified by proprietary chromatographic tech 1 * 5 µG
Catalog # BIRBORB80403-5
Supplier:  Biorbyt
IGFBP6 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing amino acids 148-240 and havi ng a molecular mass of 20 kDa including 4kDa His tag. IGFBP6 is purified by proprietary chromatographic tech 1 * 5 µG
Catalog # BIRBORB80403-5
Supplier:  Biorbyt