ContactUsLinkComponent
IGFBP3 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 2x264 amino acids and having a molecular mass of 28806 Dalton. IGFBP-3 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80401-5
Supplier:  Biorbyt
IGFBP3 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 2x264 amino acids and having a molecular mass of 28806 Dalton. IGFBP-3 is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80401-5
Supplier:  Biorbyt