MAPK3 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 399 amino acids (1-379 a.a.) and having a molecular mass of 45.2 kDa. The MAPK3 is fused to 20 amino acid His-Tag at N-terminus and purif 1 * 5 µG
Catalog # BIRBORB80798-5
Supplier: Biorbyt