ContactUsLinkComponent
ERK1/MAPK3 Recombinant is highly active form produced by phosphorylation of the purified ERK1/MAPK3 in vitro with MEK1 is non-glycos ylated polypeptide having a molecular mass of 43.6 kDa. ERK1/MAPK3 is purified by proprietary chromatographi 1 * 10 µG
Catalog # BIRBORB80796-10
Supplier:  Biorbyt
ERK1/MAPK3 Recombinant is highly active form produced by phosphorylation of the purified ERK1/MAPK3 in vitro with MEK1 is non-glycos ylated polypeptide having a molecular mass of 43.6 kDa. ERK1/MAPK3 is purified by proprietary chromatographi 1 * 10 µG
Catalog # BIRBORB80796-10
Supplier:  Biorbyt