ContactUsLinkComponent
CKS2 Human Recombinant fused to T7-Tag produced in E.coli is a single, non- glycosylated polypeptide chain containing 94 amino acids and having a molecular mass of 11 kDa. 1 * 5 µG
Catalog # BIRBORB80788-5
Supplier:  Biorbyt
CKS2 Human Recombinant fused to T7-Tag produced in E.coli is a single, non- glycosylated polypeptide chain containing 94 amino acids and having a molecular mass of 11 kDa. 1 * 5 µG
Catalog # BIRBORB80788-5
Supplier:  Biorbyt