ContactUsLinkComponent
Sonic Hedgehog Recombinant Mouse produced in E.coli is a single, non-glycosylated polypeptide chain containing 176 amino acids and h aving a molecular mass of 19.8 kDa. The Mouse Sonic Hedgehog is 99% homologous to the human gene.Cysteine at 1 * 25 µG
Catalog # BIRBORB80580-25
Supplier:  Biorbyt
Sonic Hedgehog Recombinant Mouse produced in E.coli is a single, non-glycosylated polypeptide chain containing 176 amino acids and h aving a molecular mass of 19.8 kDa. The Mouse Sonic Hedgehog is 99% homologous to the human gene.Cysteine at 1 * 25 µG
Catalog # BIRBORB80580-25
Supplier:  Biorbyt