ContactUsLinkComponent
IFNA1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 167 amino acids (24-189) and having a molecular mass of 19.5 kDa. The IFN-alpha is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80384-5
Supplier:  Biorbyt
IFNA1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 167 amino acids (24-189) and having a molecular mass of 19.5 kDa. The IFN-alpha is purified by proprietary chromatographic techniques. 1 * 5 µG
Catalog # BIRBORB80384-5
Supplier:  Biorbyt