ContactUsLinkComponent
Prolactin Ovine Antagonist Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 199 amino acids and an additional Ala at N-terminus and having a molecular mass of 23kDa. The mutant R129G is DES amino aci 1 * 50 µG
Catalog # BIRBORB80572-50
Supplier:  Biorbyt
Prolactin Ovine Antagonist Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 199 amino acids and an additional Ala at N-terminus and having a molecular mass of 23kDa. The mutant R129G is DES amino aci 1 * 50 µG
Catalog # BIRBORB80572-50
Supplier:  Biorbyt