ContactUsLinkComponent
Prolactin Ovine Antagonist Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 199 amino acids and having a molecular mass of 23kDa. Ovine Prolactin Antagonist is purified by proprietary chromatographic 1 * 10 µG
Catalog # BIRBORB80571-10
Supplier:  Biorbyt
Prolactin Ovine Antagonist Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 199 amino acids and having a molecular mass of 23kDa. Ovine Prolactin Antagonist is purified by proprietary chromatographic 1 * 10 µG
Catalog # BIRBORB80571-10
Supplier:  Biorbyt