ContactUsLinkComponent
Prolactin Rabbit Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 200 amino acids and havin g a molecular mass of 23007 Dalton. The Porlactin is purified by proprietary chromatographic techniques. 1 * 50 µG
Catalog # BIRBORB80563-50
Supplier:  Biorbyt
Prolactin Rabbit Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 200 amino acids and havin g a molecular mass of 23007 Dalton. The Porlactin is purified by proprietary chromatographic techniques. 1 * 50 µG
Catalog # BIRBORB80563-50
Supplier:  Biorbyt