ContactUsLinkComponent
HGF-B Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 234 amino acids fragment (495 -728) having molecular weight of 34kDa and fused with 4.5kDa amino-terminal hexahistidine tag. The HGF-B is 1 * 10 µG
Catalog # BIRBORB80382-10
Supplier:  Biorbyt
HGF-B Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 234 amino acids fragment (495 -728) having molecular weight of 34kDa and fused with 4.5kDa amino-terminal hexahistidine tag. The HGF-B is 1 * 10 µG
Catalog # BIRBORB80382-10
Supplier:  Biorbyt