ContactUsLinkComponent
GMCSF Human Recombinant produced in insect cells is a single, glycosylated, polypeptide chain containing 127 amino acids (18-144) an d having a molecular mass of 14.6kDa. GMCSF is fused to C-terminal His -tag (6x His) and purified by proprie 1 * 2 µG
Catalog # BIRBORB80372-2
Supplier:  Biorbyt
GMCSF Human Recombinant produced in insect cells is a single, glycosylated, polypeptide chain containing 127 amino acids (18-144) an d having a molecular mass of 14.6kDa. GMCSF is fused to C-terminal His -tag (6x His) and purified by proprie 1 * 2 µG
Catalog # BIRBORB80372-2
Supplier:  Biorbyt