ContactUsLinkComponent
Placental Lactogen Bovine Recombinant, is a single polypeptide chain containing 199 amino acids and an additional Ala at the N-termi nus having a molecular mass of 23 kDa. Placental Lactogen Recombinant is purified by proprietary chromatogra 1 * 1 mg
Catalog # BIRBORB80553-1
Supplier:  Biorbyt
Placental Lactogen Bovine Recombinant, is a single polypeptide chain containing 199 amino acids and an additional Ala at the N-termi nus having a molecular mass of 23 kDa. Placental Lactogen Recombinant is purified by proprietary chromatogra 1 * 1 mg
Catalog # BIRBORB80553-1
Supplier:  Biorbyt