ContactUsLinkComponent
Prolactin Ovine Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 199 amino acids and having a molecular mass of 23 kDa. The Prolactin is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80562-10
Supplier:  Biorbyt
Prolactin Ovine Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 199 amino acids and having a molecular mass of 23 kDa. The Prolactin is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80562-10
Supplier:  Biorbyt