ContactUsLinkComponent
Somatotropin Ovine Antagonist Recombinant G119R produced in E.coli is a single, non-glycosylated polypeptide chain containing 191 am ino acids and having a molecular mass of 22 kDa. The Somatotropin Ovine Antagonist Recombinant is purified b 1 * 1 mg
  BIRBORB80368-1
 :  Biorbyt
Somatotropin Ovine Antagonist Recombinant G119R produced in E.coli is a single, non-glycosylated polypeptide chain containing 191 am ino acids and having a molecular mass of 22 kDa. The Somatotropin Ovine Antagonist Recombinant is purified b 1 * 1 mg
  BIRBORB80368-1
 :  Biorbyt