ContactUsLinkComponent
Somatotropin Chicken Antagonist Recombinant mutein G119R produced in E.coli is a single, non-glycosylated polypeptide chain containi ng 191 amino acids with an additional Ala at the N-terminus and having a molecular mass of 22.3 kDa. The Chi 1 * 100 µG
Catalog # BIRBORB80367-100
Supplier:  Biorbyt
Somatotropin Chicken Antagonist Recombinant mutein G119R produced in E.coli is a single, non-glycosylated polypeptide chain containi ng 191 amino acids with an additional Ala at the N-terminus and having a molecular mass of 22.3 kDa. The Chi 1 * 100 µG
Catalog # BIRBORB80367-100
Supplier:  Biorbyt