ContactUsLinkComponent
Porcine-Somatotropin Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 190 amino acids and having a molecular mass of 21730 Dalton. Growth Hormone is purified by proprietary chromatographic technique 1 * 1 mg
Catalog # BIRBORB80366-1
Supplier:  Biorbyt
Porcine-Somatotropin Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 190 amino acids and having a molecular mass of 21730 Dalton. Growth Hormone is purified by proprietary chromatographic technique 1 * 1 mg
Catalog # BIRBORB80366-1
Supplier:  Biorbyt