ContactUsLinkComponent
Growth Hormone Rat Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 190 amino acids and ha ving a molecular mass of 21810 Dalton. GH is purified by proprietary chromatographic techniques. 1 * 100 µG
Catalog # BIRBORB80360-100
Supplier:  Biorbyt
Growth Hormone Rat Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 190 amino acids and ha ving a molecular mass of 21810 Dalton. GH is purified by proprietary chromatographic techniques. 1 * 100 µG
Catalog # BIRBORB80360-100
Supplier:  Biorbyt