ContactUsLinkComponent
Growth Hormone Mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 191 amino acids and having a molecular mass of 22 kDa. Mouse GH is purified by proprietary chromatographic techniques. 1 * 200 µG
Catalog # BIRBORB80359-200
Supplier:  Biorbyt
Growth Hormone Mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 191 amino acids and having a molecular mass of 22 kDa. Mouse GH is purified by proprietary chromatographic techniques. 1 * 200 µG
Catalog # BIRBORB80359-200
Supplier:  Biorbyt