Growth Hormone Mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 191 amino acids and having a molecular mass of 22 kDa. Mouse GH is purified by proprietary chromatographic techniques. 1 * 100 µG
Catalog # BIRBORB80359-100
Supplier: Biorbyt