ContactUsLinkComponent
c-erbB2 Human Recombinant encoding 537-636 amino acids expressed in E.colic-erbB2 is purified by proprietary chromatographic techniq ues. 1 * 10 µG
Catalog # BIRBORB80781-10
Supplier:  Biorbyt
c-erbB2 Human Recombinant encoding 537-636 amino acids expressed in E.colic-erbB2 is purified by proprietary chromatographic techniq ues. 1 * 10 µG
Catalog # BIRBORB80781-10
Supplier:  Biorbyt