ContactUsLinkComponent
Growth Hormone Bovine Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 191 amino acids and having a molecular mass of 21.8 kDa. The GH Bovine Recombinant is purified by proprietary chromatographic te 1 * 100 µG
Catalog # BIRBORB80358-100
Supplier:  Biorbyt
Growth Hormone Bovine Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 191 amino acids and having a molecular mass of 21.8 kDa. The GH Bovine Recombinant is purified by proprietary chromatographic te 1 * 100 µG
Catalog # BIRBORB80358-100
Supplier:  Biorbyt