Growth Hormone Ovine Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 200 amino acids and h aving a molecular mass of 22015 Dalton. The GH Ovine Recombinant is purified by proprietary chromatographic 1 * 1 mg
Catalog # BIRBORB80356-1
Supplier: Biorbyt