ContactUsLinkComponent
Granulocyte Colony Stimulating Factor Human Recombinant produced in E.coli is a single, glycosylated, polypeptide chain containing 1 74 amino acids and having a molecular mass of approximately 18 kDa.GCSF is purified by proprietary chromatog 1 * 10 µG
Catalog # BIRBORB80346-10
Supplier:  Biorbyt
Granulocyte Colony Stimulating Factor Human Recombinant produced in E.coli is a single, glycosylated, polypeptide chain containing 1 74 amino acids and having a molecular mass of approximately 18 kDa.GCSF is purified by proprietary chromatog 1 * 10 µG
Catalog # BIRBORB80346-10
Supplier:  Biorbyt