ContactUsLinkComponent
Granulocyte Colony Stimulating Factor-His Tag Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 174 amino acids, fragment (31-204) and having a molecular mass of 23.19 kDa with an amino-termina 1 * 2 µG
Catalog # BIRBORB80345-2
Supplier:  Biorbyt
Granulocyte Colony Stimulating Factor-His Tag Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 174 amino acids, fragment (31-204) and having a molecular mass of 23.19 kDa with an amino-termina 1 * 2 µG
Catalog # BIRBORB80345-2
Supplier:  Biorbyt