ContactUsLinkComponent
OSTF1 Human RECOMB produced in E.coli is monomeric, non-glycosylated, polypeptide chain containing 222 amino acids (1-214) and havin g a molecular mass of 25.1 kDa. The OSTF1 is fused to amino acid His Tag at C-terminus purified by proprieta 1 * 10 µG
Catalog # BIRBORB80537-10
Supplier:  Biorbyt
OSTF1 Human RECOMB produced in E.coli is monomeric, non-glycosylated, polypeptide chain containing 222 amino acids (1-214) and havin g a molecular mass of 25.1 kDa. The OSTF1 is fused to amino acid His Tag at C-terminus purified by proprieta 1 * 10 µG
Catalog # BIRBORB80537-10
Supplier:  Biorbyt