Recombinant Human Neuregulin-1 beta produced in E.coli is a single, non-glycosylated, polypeptide chain containing 61 amino acids an d having total molecular mass of 7055 Dalton. NRG-1 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80530-10
Supplier: Biorbyt