ContactUsLinkComponent
GDF10 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 111 amino acids (369-478 a.a. ) and having total molecular mass of 12.5 kDa. GDF10 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80340-20
Supplier:  Biorbyt
GDF10 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 111 amino acids (369-478 a.a. ) and having total molecular mass of 12.5 kDa. GDF10 is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80340-20
Supplier:  Biorbyt