ContactUsLinkComponent
Galectin 4 produced in E.coli is a single, non-glycosylated polypeptide chain containing 343 amino acids (1-323 a.a.) and having a m olecular mass of 38.1kDa.Galectin 4 is fused to 20 amino acid His-Tag at N-terminus and purified by propriet 1 * 25 µG
  BIRBORB80333-25
 :  Biorbyt
Galectin 4 produced in E.coli is a single, non-glycosylated polypeptide chain containing 343 amino acids (1-323 a.a.) and having a m olecular mass of 38.1kDa.Galectin 4 is fused to 20 amino acid His-Tag at N-terminus and purified by propriet 1 * 25 µG
  BIRBORB80333-25
 :  Biorbyt