ContactUsLinkComponent
AK3L1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 259 amino acids (1-223) and h aving a molecular mass of 29.3 kDa. The AK3L1 is fused with 35 amino acid His-tag at N-terminus.AK3L1 is pur 1 * 5 µG
Catalog # BIRBORB80771-5
Supplier:  Biorbyt
AK3L1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 259 amino acids (1-223) and h aving a molecular mass of 29.3 kDa. The AK3L1 is fused with 35 amino acid His-tag at N-terminus.AK3L1 is pur 1 * 5 µG
Catalog # BIRBORB80771-5
Supplier:  Biorbyt