Triptorelin C64H82N18O13, Pyr-His-Trp-Ser-Tyr-D-Trp-Leu-Arg-Pro-Gly-NH2 is synthetic analogue of gonadorelin (GnRH). As result of th e substitution of the 6th amino acid residue in the native molecule, the agonistic effect is more pronounced 1 * 5 mg
Catalog # BIRBORB80765-5
Supplier: Biorbyt