ContactUsLinkComponent
AURKA Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 423 amino acids (1-403) and h aving a molecular mass of 47.9kDa. AURKA is fused to 20 a.a. His-Tag at N-terminus and purified by proprieta 1 * 10 µG
Catalog # BIRBORB80774-10
Supplier:  Biorbyt
AURKA Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 423 amino acids (1-403) and h aving a molecular mass of 47.9kDa. AURKA is fused to 20 a.a. His-Tag at N-terminus and purified by proprieta 1 * 10 µG
Catalog # BIRBORB80774-10
Supplier:  Biorbyt
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.