ContactUsLinkComponent
Follistatin Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 288 amino acids and havi ng total molecular mass of 31.5kDa.The FST is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80327-20
Supplier:  Biorbyt
Follistatin Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 288 amino acids and havi ng total molecular mass of 31.5kDa.The FST is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80327-20
Supplier:  Biorbyt